Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117994.4 | NP_567564.1 | 4 - 5 | 646 | 68 | |
GenBank mRNA | AY099636.1 | - | 4 - 5 | 546 | 68 | |
KJ676982.1 | - | 4 - 5 | 479 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |