Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343715.1 | NP_001330213.1 | 6 - 7 | 936 | 73 | |
NM_147883.6 | NP_680188.1 | 6 - 7 | 936 | 73 | ||
GenBank mRNA | BT014965.1 | - | 4 - 5 | 734 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |