Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105392.5 | NP_176892.1 | 7 - 8 | 3554 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |