Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198420.1 | NP_001185349.1 | 2 - 3 | 436 | 86 | |
NM_001334339.1 | NP_001320607.1 | 2 - 3 | 414 | 86 | ||
NM_105471.4 | NP_176968.1 | 2 - 3 | 439 | 86 | ||
GenBank mRNA | AF370221.1 | - | 2 - 3 | 279 | 86 | |
AK226564.1 | - | 256 | 86 | |||
AY099552.1 | - | 2 - 3 | 271 | 86 | ||
AY113871.1 | - | 2 - 3 | 214 | 86 | ||
BT001237.1 | - | 2 - 3 | 214 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |