Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105557.6 | NP_564946.1 | 4 - 5 | 890 | 84 | |
GenBank mRNA | BT030352.1 | - | 4 - 5 | 701 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |