Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105604.2 | NP_177096.1 | 1 - 2 | 307 | 163 | |
GenBank mRNA | AK117860.1 | - | 1 - 2 | 181 | 163 | |
BT005306.1 | - | 1 - 2 | 165 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |