Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105699.4 | NP_177188.1 | 7 - 8 | 1199 | 109 | |
GenBank mRNA | AF375409.1 | - | 7 - 8 | 1016 | 109 | |
AJ251297.1 | - | 7 - 8 | 988 | 109 | ||
BT020360.1 | - | 7 - 8 | 956 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |