Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334497.1 | NP_001321997.1 | 1 | 1795 | 162 | |
NM_105808.2 | NP_177295.1 | 1 | 1743 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |