Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198445.1 | NP_001185374.1 | 6 - 7 | 802 | 95 | |
NM_105840.4 | NP_565026.1 | 6 - 7 | 787 | 95 | ||
GenBank mRNA | AY046014.1 | - | 6 - 7 | 708 | 95 | |
AY133819.1 | - | 5 - 6 | 439 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |