Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334580.1 | NP_001319372.1 | 6 - 7 | 738 | 78 | |
NM_001334581.1 | NP_001323094.1 | 6 - 7 | 737 | 78 | ||
NM_001334582.1 | NP_001323093.1 | 6 - 7 | 737 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |