Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL09 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344150.1 | NP_001329981.1 | 19 - 20 | 2163 | 73 | |
NM_001344151.1 | NP_001329982.1 | 19 - 20 | 2163 | 73 | ||
NM_001344152.1 | NP_001329979.1 | 18 - 19 | 2234 | 73 | ||
NM_001344153.1 | NP_001329980.1 | 18 - 19 | 2080 | 73 | ||
NM_123045.4 | NP_198503.3 | 18 - 19 | 2083 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |