Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337509.1 | NP_001326045.1 | 19 - 20 | 2314 | 66 | |
NM_111295.4 | NP_187074.1 | 25 - 26 | 2914 | 66 | ||
GenBank mRNA | AF441079.1 | - | 25 - 26 | 2832 | 66 | |
AK226884.1 | - | 25 - 26 | 2857 | 66 | ||
AY090938.1 | - | 25 - 26 | 2853 | 66 | ||
AY117340.1 | - | 25 - 26 | 2703 | 66 | ||
KJ138841.1 | - | 25 - 26 | 2703 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |