Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111295.4 | NP_187074.1 | 2 - 3 | 505 | 97 | |
GenBank mRNA | AF441079.1 | - | 2 - 3 | 423 | 97 | |
AK226884.1 | - | 2 - 3 | 448 | 97 | ||
AY090938.1 | - | 2 - 3 | 444 | 97 | ||
AY117340.1 | - | 2 - 3 | 294 | 97 | ||
KJ138841.1 | - | 2 - 3 | 294 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |