Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339194.1 | NP_001327790.1 | 9 - 10 | 1492 | 87 | |
NM_001339195.1 | NP_001327789.1 | 8 - 9 | 1399 | 87 | ||
NM_001339196.1 | NP_001327788.1 | 8 - 9 | 1599 | 87 | ||
NM_001339197.1 | NP_001327787.1 | 9 - 10 | 1492 | 87 | ||
NM_114382.5 | NP_190099.3 | 8 - 9 | 1380 | 87 | ||
GenBank mRNA | AB247155.1 | - | 8 - 9 | 1340 | 87 | |
DQ508794.2 | - | 8 - 9 | 1340 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |