Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124823.2 | NP_001118295.1 | 4 - 5 | 726 | 98 | |
NM_001335364.1 | NP_001324918.1 | 4 - 5 | 656 | 98 | ||
NM_001335365.1 | NP_001324917.1 | 4 - 5 | 620 | 98 | ||
NM_126831.4 | NP_178872.2 | 4 - 5 | 738 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |