Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035893.1 | NP_001030970.1 | 6 - 7 | 566 | 70 | |
NM_100396.4 | NP_172010.1 | 5 - 6 | 793 | 70 | ||
NM_202032.2 | NP_973761.1 | 6 - 7 | 566 | 70 | ||
GenBank mRNA | AY050379.1 | - | 5 - 6 | 653 | 70 | |
L13922.1 | - | 5 - 6 | 632 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |