Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102139.2 | NP_173705.1 | 1 - 2 | 561 | 92 | |
NM_202161.1 | NP_973890.1 | 1 - 2 | 551 | 92 | ||
GenBank mRNA | AF042334.1 | - | 1 - 2 | 415 | 92 | |
AF395062.1 | - | 1 - 2 | 395 | 92 | ||
BT029486.1 | - | 1 - 2 | 395 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |