Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLP31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111410.4 | NP_187188.2 | 1 | 2723 | 105 | |
GenBank mRNA | AY090373.1 | - | 1 | 2572 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |