Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337604.1 | NP_001319482.1 | 2 - 3 | 350 | 89 | |
NM_111446.2 | NP_187223.1 | 2 - 3 | 285 | 89 | ||
GenBank mRNA | DQ446636.1 | - | 2 - 3 | 265 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |