Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337604.1 | NP_001319482.1 | 5 - 6 | 766 | 60 | |
NM_111446.2 | NP_187223.1 | 5 - 6 | 701 | 60 | ||
GenBank mRNA | DQ446636.1 | - | 5 - 6 | 681 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |