Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LON4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337610.1 | NP_001325548.1 | 9 - 10 | 1064 | 76 | |
NM_111453.1 | NP_566259.1 | 9 - 10 | 1011 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |