Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCI2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111462.2 | NP_187239.1 | 2 - 3 | 616 | 80 | |
GenBank mRNA | AF122005.1 | - | 2 - 3 | 260 | 80 | |
AY037244.1 | - | 2 - 3 | 263 | 80 | ||
AY039859.1 | - | 2 - 3 | 263 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |