Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIP7;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337632.1 | NP_001327693.1 | 3 - 4 | 586 | 62 | |
NM_001337633.1 | NP_001327692.1 | 2 - 3 | 406 | 62 | ||
NM_111485.3 | NP_566271.1 | 4 - 5 | 895 | 62 | ||
GenBank mRNA | DQ446638.1 | - | 4 - 5 | 633 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |