Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337650.1 | NP_001327378.1 | 11 - 12 | 1202 | 87 | |
NM_001337651.1 | NP_001327379.1 | 11 - 12 | 1202 | 87 | ||
NM_001337652.1 | NP_001327377.1 | 10 - 11 | 1132 | 87 | ||
NM_111512.3 | NP_187288.2 | 11 - 12 | 1202 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |