Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111513.4 | NP_187289.1 | 1 - 2 | 601 | 151 | |
GenBank mRNA | AF487270.1 | - | 1 - 2 | 290 | 151 | |
BT026465.1 | - | 1 - 2 | 290 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |