Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111534.4 | NP_187310.1 | 5 - 6 | 849 | 132 | |
GenBank mRNA | AY136356.1 | - | 5 - 6 | 728 | 132 | |
BT010374.1 | - | 5 - 6 | 569 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |