Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111566.4 | NP_187342.1 | 10 - 11 | 1257 | 132 | |
GenBank mRNA | AF123254.1 | - | 10 - 11 | 1066 | 132 | |
AY062621.1 | - | 10 - 11 | 1123 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |