Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036629.2 | NP_001031706.1 | 8 - 9 | 1175 | 147 | |
NM_118530.5 | NP_194129.1 | 8 - 9 | 1230 | 147 | ||
GenBank mRNA | AF082176.1 | - | 8 - 9 | 1068 | 147 | |
AF344312.1 | - | 8 - 9 | 781 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |