Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118536.4 | NP_194135.1 | 10 - 11 | 2003 | 94 | |
GenBank mRNA | AK118407.1 | - | 10 - 11 | 1860 | 94 | |
BT010732.1 | - | 10 - 11 | 1801 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |