Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341655.1 | NP_001328763.1 | 11 - 12 | 1420 | 84 | |
NM_118558.4 | NP_567697.1 | 12 - 13 | 1337 | 84 | ||
GenBank mRNA | AF369574.1 | - | 12 - 13 | 1301 | 84 | |
AK221618.1 | - | 12 - 13 | 1290 | 84 | ||
BT026373.1 | - | 12 - 13 | 1271 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |