Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341655.1 | NP_001328763.1 | 5 - 6 | 633 | 73 | |
NM_118558.4 | NP_567697.1 | 5 - 6 | 633 | 73 | ||
GenBank mRNA | AF369574.1 | - | 5 - 6 | 597 | 73 | |
AK221618.1 | - | 5 - 6 | 586 | 73 | ||
BT026373.1 | - | 5 - 6 | 567 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |