Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341655.1 | NP_001328763.1 | 7 - 8 | 816 | 138 | |
NM_118558.4 | NP_567697.1 | 7 - 8 | 816 | 138 | ||
GenBank mRNA | AF369574.1 | - | 7 - 8 | 780 | 138 | |
AK221618.1 | - | 7 - 8 | 769 | 138 | ||
BT026373.1 | - | 7 - 8 | 750 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |