Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVA22D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203901.1 | NP_001190830.1 | 2 - 3 | 195 | 66 | |
NM_118628.4 | NP_567713.1 | 2 - 3 | 275 | 66 | ||
GenBank mRNA | AF141662.1 | - | 2 - 3 | 194 | 66 | |
AY059750.1 | - | 2 - 3 | 194 | 66 | ||
AY086850.1 | - | 2 - 3 | 194 | 66 | ||
AY113978.1 | - | 2 - 3 | 71 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |