Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341722.1 | NP_001320059.1 | 26 - 27 | 3383 | 99 | |
GenBank mRNA | GQ148553.1 | - | 26 - 27 | 3290 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |