Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036643.2 | NP_001031720.1 | 7 - 8 | 2235 | 84 | |
NM_001341767.1 | NP_001329827.1 | 6 - 7 | 2193 | 84 | ||
NM_001341768.1 | NP_001329828.1 | 6 - 7 | 2193 | 84 | ||
NM_118721.3 | NP_567733.1 | 7 - 8 | 2514 | 84 | ||
NM_202889.3 | NP_974618.2 | 7 - 8 | 2315 | 84 | ||
GenBank mRNA | AF360298.1 | - | 7 - 8 | 2330 | 84 | |
AK319148.1 | - | 6 - 7 | 2684 | 84 | ||
AY133866.1 | - | 6 - 7 | 2193 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |