Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118733.4 | NP_194330.1 | 4 - 5 | 1227 | 126 | |
GenBank mRNA | AY056288.1 | - | 4 - 5 | 1050 | 126 | |
AY099659.1 | - | 4 - 5 | 1044 | 126 | ||
AY114064.1 | - | 4 - 5 | 1016 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |