Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118867.2 | NP_194463.1 | 2 - 3 | 898 | 107 | |
GenBank mRNA | AB493704.1 | - | 2 - 3 | 763 | 107 | |
DQ446873.1 | - | 2 - 3 | 763 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |