Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIIIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341848.1 | NP_001328308.1 | 18 - 19 | 2348 | 127 | |
NM_001341849.1 | NP_001328309.1 | 17 - 18 | 2287 | 127 | ||
NM_001341850.1 | NP_001328307.1 | 17 - 18 | 2364 | 127 | ||
NM_118871.6 | NP_194467.5 | 16 - 17 | 2155 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |