Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATKRS-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112003.4 | NP_187777.1 | 12 - 13 | 1694 | 68 | |
GenBank mRNA | AF125574.1 | - | 12 - 13 | 1677 | 68 | |
AY099551.1 | - | 12 - 13 | 1655 | 68 | ||
BT002123.1 | - | 12 - 13 | 1591 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |