Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337982.1 | NP_001327319.1 | 10 - 11 | 1242 | 68 | |
NM_001337983.1 | NP_001327318.1 | 9 - 10 | 1013 | 68 | ||
NM_112058.4 | NP_566414.3 | 11 - 12 | 1169 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |