Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATMND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085000.2 | NP_001078469.1 | 8 - 9 | 748 | 79 | |
NM_119061.4 | NP_194646.2 | 8 - 9 | 653 | 79 | ||
GenBank mRNA | AK117713.1 | - | 8 - 9 | 602 | 79 | |
AM162278.1 | - | 7 - 8 | 504 | 79 | ||
BT005435.1 | - | 7 - 8 | 504 | 79 | ||
DQ248000.1 | - | 8 - 9 | 625 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |