Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119078.5 | NP_194662.2 | 6 - 7 | 995 | 83 | |
GenBank mRNA | AY063817.1 | - | 6 - 7 | 728 | 83 | |
AY096457.1 | - | 6 - 7 | 611 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |