Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119079.4 | NP_194663.1 | 2 - 3 | 653 | 118 | |
GenBank mRNA | AY085179.1 | - | 2 - 3 | 342 | 118 | |
BT024816.1 | - | 2 - 3 | 273 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |