Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACHT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119112.3 | NP_567831.1 | 2 - 3 | 676 | 64 | |
NM_179138.3 | NP_849469.1 | 2 - 3 | 672 | 64 | ||
GenBank mRNA | AF370616.1 | - | 2 - 3 | 428 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |