Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203938.2 | NP_001190867.1 | 6 - 7 | 1470 | 93 | |
NM_119140.2 | NP_194723.1 | 6 - 7 | 1446 | 93 | ||
GenBank mRNA | L21991.1 | - | 6 - 7 | 1361 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |