Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342010.1 | NP_001320091.1 | 6 - 7 | 584 | 64 | |
NM_001342011.1 | NP_001320092.1 | 4 - 5 | 768 | 64 | ||
GenBank mRNA | AY142529.1 | - | 4 - 5 | 370 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |