Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119243.4 | NP_194824.1 | 2 - 3 | 272 | 86 | |
GenBank mRNA | AY045621.1 | - | 2 - 3 | 197 | 86 | |
AY058078.1 | - | 2 - 3 | 172 | 86 | ||
AY058852.1 | - | 2 - 3 | 202 | 86 | ||
AY079039.1 | - | 2 - 3 | 99 | 86 | ||
AY088002.1 | - | 2 - 3 | 214 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |