Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345081.1 | NP_001330550.1 | 16 - 17 | 1512 | 150 | |
NM_124806.2 | NP_200237.1 | 18 - 19 | 1698 | 150 | ||
GenBank mRNA | AJ243822.1 | - | 18 - 19 | 1667 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |