Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345094.1 | NP_001330580.1 | 14 - 15 | 2109 | 79 | |
NM_001345095.1 | NP_001330581.1 | 12 - 13 | 1733 | 79 | ||
NM_124848.3 | NP_568811.1 | 14 - 15 | 2228 | 79 | ||
GenBank mRNA | AK228138.1 | - | 14 - 15 | 2159 | 79 | |
BT003156.1 | - | 13 - 14 | 1880 | 79 | ||
D21138.1 | - | 14 - 15 | 2130 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |