Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345121.1 | NP_001330655.1 | 6 - 7 | 1218 | 66 | |
NM_124908.4 | NP_200337.1 | 7 - 8 | 1093 | 66 | ||
GenBank mRNA | BT024716.1 | - | 7 - 8 | 865 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |